Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABEP2 Monoclonal Antibody (2B2A6)
GREENER_CHOICE

Catalog No. FERMA548783
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548783 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548783 Supplier Invitrogen™ Supplier No. MA548783
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RABEP2 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA

RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABEP2
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2B2A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RABEP2
Gene Accession No. Q9H5N1
Gene Alias 2610011A08Rik; AI462746; fos-related antigen; Fra; MP13; MP13 protein; Rab GTPase-binding effector protein 2; rabaptin, RAB GTPase binding effector protein 2; Rabaptin-5 beta; rabaptin-5beta; RABEP2; RABPT5B
Gene Symbols RABEP2
Host Species Mouse
Immunogen Purified recombinant fragment of human RABEP2 (AA: 10-200) expressed in HEK293-6e cells supernatant.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.