Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABEP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561562
Description
Immunogen sequence: LKAKLLRAEE LIQEIQRRPR HAPSLHGSTE LLPLSRDPSP PLEPLEELSG DGGPAAEAFA HNCDDSASIS SFSLGGGVGS SSSLPQSRQG LSPE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.
Specifications
RABEP2 | |
Polyclonal | |
Unconjugated | |
RABEP2 | |
2610011A08Rik; AI462746; fos-related antigen; Fra; MP13; MP13 protein; Rab GTPase-binding effector protein 2; rabaptin, RAB GTPase binding effector protein 2; Rabaptin-5 beta; rabaptin-5beta; RABEP2; RABPT5B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5N1 | |
RABEP2 | |
Recombinant protein corresponding to Human RABEP2. Recombinant protein control fragment (Product #RP-98772). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction