Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABEPK/p40 Mouse anti-Human, Mouse, Rat, Clone: 1E11A3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RABEPK/p40 |
---|---|
Clone | 1E11A3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7040
![]() ![]() |
Proteintech Group Inc
666221IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.Specifications
RABEPK/p40 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
40 kDa Rab9 effector protein, p40, RAB9P40, RABEPK | |
Rabepk | |
IgG1 | |
Protein G | |
Antibody |
1E11A3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q4V8F4, Q7Z6M1, Q8VCH5 | |
10244, 227746, 296649 | |
RABEPK/p40 Fusion Protein Ag7796 | |
Primary | |
-20°C | |
RABEPK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title