Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABEPK/p40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726118 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6118 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6118 Supplier Proteintech Group Inc Supplier No. 102132AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rab9 effector required for endosome to trans-Golgi network (TGN) transport.

Specifications

Antigen RABEPK/p40
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RABEPK
Gene Accession No. Q4V8F4, Q7Z6M1, Q8VCH5
Gene Alias 40 kDa Rab9 effector protein, p40, RAB9P40, RABEPK
Gene Symbols Rabepk
Host Species Rabbit
Immunogen RABEPK/p40 Fusion Protein Ag0285
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10244, 227746, 296649
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.