Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Rabex-5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544268
Description
Peptide sequence: MSLKSERRGI HVDQSDLLCK KGCGYYGNPA WQGFCSKCWR EEYHKARQKQ Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.
Specifications
Rabex-5 | |
Polyclonal | |
Unconjugated | |
RABGEF1 | |
BTB/POZ domain-containing protein KCTD7; RAB guanine nucleotide exchange factor (GEF) 1; RAB guanine nucleotide exchange factor 1; Rab5 exchange factor; rab5 GDP/GTP exchange factor; Rab5 GDP/GTP exchange factor, Rabex5; Rab5 guanine nucleotide exchange factor Rabex-5; Rab5ef; Rabaptin-5-associated exchange factor for Rab5; Rabex5; rabex-5; RABEX5 protein; RABGEF1; RAP1; Ras negative regulator Rabex-5; Ras negative regulator Rabex-5/Rin2; Rin2; zgc:65944 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27342 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJ41 | |
RABGEF1 | |
Synthetic peptide directed towards the N-terminal of human RABGEF1 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction