Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabex5 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297267AF700
Description
Rabex5 Polyclonal specifically detects Rabex5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Rabex5 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
FLJ32302, RAB guanine nucleotide exchange factor (GEF) 1, rab5 GDP/GTP exchange factor, Rabaptin-5-associated exchange factor for Rab5, Rabex-5, RABEX5rabex-5, RAP1 | |
Produced in rabbits immunized with E. coli-derived Human Rabex5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
27342 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction