Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABGAP1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244285DL594
Description
RABGAP1 Polyclonal antibody specifically detects RABGAP1 in Human samples. It is validated for Western BlotSpecifications
RABGAP1 | |
Polyclonal | |
Western Blot | |
DKFZp586D2123, GAP and centrosome-associated protein, GAPCENA, RAB GTPase activating protein 1, rab GTPase-activating protein 1, rab6 GTPase activating protein (GAP and centrosome-associated), Rab6 GTPase activating protein, GAPCenA, Rab6 GTPase-activating protein GAPCenA, RP11-123N4.2, TBC1 domain family, member 11, TBC1D11 | |
The epitope recognized by this antibody maps to a region between residue 25 to 75 of human Rab GTPase-activating protein 1 using the numbering given in entry Q9Y3P9.3 (GeneID 23637). | |
0.1 mL | |
Cell Cycle and Replication | |
23637 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction