Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABGAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567235 Supplier Invitrogen™ Supplier No. PA567235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKQQTANKVEI EKIRQKVDDC ERCREFFNKE GRVKGISSTK EVLDEDTDEE KETLKN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.

RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABGAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RABGAP1
Gene Accession No. Q9Y3P9
Gene Alias G protein-coupled receptor 21; GAP and centrosome-associated protein; GAPCENA; Gpr21; HSPC094; Kiaa4104; mKIAA4104; RAB GTPase activating protein 1; rab GTPase-activating protein 1; rab6 GTPase activating protein (GAP and centrosome-associated); Rab6 GTPase activating protein, GAPCenA; rab6 GTPase-activating protein GAPCenA; RABGAP1; RP11-123N4.2; TBC1 domain family, member 11; TBC1D11
Gene Symbols RABGAP1
Host Species Rabbit
Immunogen Recombinant Human RABGAP1. Recombinant protein control fragment (Product #RP-105559).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.