Learn More
Invitrogen™ RABGAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110263
Description
Immunogen sequence: QQEDPIERFE RENRRLQEAN MRLEQENDDL AHELVTSKIA LRKDLDNAEE KADALNKELL MTKQKLIDAE EEKRRLEEES AQLKEMCRRE LDKAESEIKK NSSIIGDYKQ ICSQLSERLE KQQTANKVEI EKIRQKVDDC ERCREFFNKE GRVKGISSTK EVLDEDTDEE KETLKNQLRE MELELAQTKL QLVEAECKIQ DLEHHLGLAL NEVQAAKKTW FNRTLSSIKT ATGVQGKETC.
RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.
Specifications
| RABGAP1 | |
| Polyclonal | |
| Unconjugated | |
| RABGAP1 | |
| G protein-coupled receptor 21; GAP and centrosome-associated protein; GAPCENA; Gpr21; HSPC094; Kiaa4104; mKIAA4104; RAB GTPase activating protein 1; rab GTPase-activating protein 1; rab6 GTPase activating protein (GAP and centrosome-associated); Rab6 GTPase activating protein, GAPCenA; rab6 GTPase-activating protein GAPCenA; RABGAP1; RP11-123N4.2; TBC1 domain family, member 11; TBC1D11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 227800, 23637 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A2AWA9, Q9Y3P9 | |
| RABGAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 830-1069 of human RABGAP1 (NP_036329.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.