Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABGAP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569012
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9012 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9012 Supplier Bethyl Laboratories, Inc Supplier No. A304913AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan

RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.

Specifications

Antigen RABGAP1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene RABGAP1
Gene Accession No. Q9Y3P9
Gene Alias GAPCENA, RP11-123N4.2, TBC1D11, GAP and centrosome-associated protein, Rab6 GTPase activating protein, GAPCenA, TBC1 domain family, member 11, rab GTPase-activating protein 1, rab6 GTPase activating protein (GAP and centrosome-associated), rab6 GTPase-activating protein GAPCenA
Gene Symbols RABGAP1
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23637
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.