Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABGGTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569573 Supplier Invitrogen™ Supplier No. PA569573
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Catalyzes the transfer of a geranylgeranyl moiety from geranylgeranyl diphosphate to both cysteines of Rab proteins with the C-terminal sequence -XXCC, -XCXC and -CCXX, such as RAB1A, RAB3A, RAB5A and RAB7A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABGGTA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RABGGTA
Gene Accession No. Q92696
Gene Alias Geranylgeranyl transferase type II subunit alpha; geranylgeranyl transferase type-2 subunit alpha; Ggta; gm; gunmetal; protein prenyltransferase alpha subunit repeat containing 3; PTAR3; Rab geranylgeranyl transferase, a subunit; Rab geranylgeranyltransferase alpha; Rab geranylgeranyltransferase alpha subunit; Rab geranylgeranyltransferase subunit alpha; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; rab GG transferase alpha; rab GGTase alpha; Rabggta
Gene Symbols RABGGTA
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human RABGGTA.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5875
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.