Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABGGTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598767
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598767 Supplier Thermo Scientific Supplier No. PA598767
Add to Cart
Add to Cart

RABGGTA Polyclonal Antibody

RABGGTA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen RABGGTA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RABGGTA
Gene Accession No. Q92696, Q9JHK4
Gene Alias Geranylgeranyl transferase type II subunit alpha; geranylgeranyl transferase type-2 subunit alpha; Ggta; gm; gunmetal; protein prenyltransferase alpha subunit repeat containing 3; PTAR3; Rab geranylgeranyl transferase, a subunit; Rab geranylgeranyltransferase alpha; Rab geranylgeranyltransferase alpha subunit; Rab geranylgeranyltransferase subunit alpha; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; rab GG transferase alpha; rab GGTase alpha; Rabggta
Gene Symbols RABGGTA
Host Species Rabbit
Immunogen Recombinant Human Geranylgeranyl transferase type-2 subunit alpha protein (1-200AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56187, 5875
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.