Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABGGTB Antibody, Novus Biologicals™
SDP

Catalog No. NB406328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406328 25 μL
NBP185957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406328 Supplier Novus Biologicals Supplier No. NBP18595725UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RABGGTB Polyclonal specifically detects RABGGTB in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RABGGTB
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 2.5.1.60, Geranylgeranyl transferase type II subunit beta, geranylgeranyl transferase type-2 subunit beta, GGTase-II-beta, GGTB, Rab geranylgeranyltransferase subunit beta, Rab geranyl-geranyltransferase subunit beta, Rab geranylgeranyltransferase, beta subunit, Rab GG transferase beta, Rab GGTase beta, Type II protein geranyl-geranyltransferase subunit beta
Gene Symbols RABGGTB
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MGTPQKDVIIKSDAPDTLLLEKHADYIASYGSKKDDYEYCMSEYLRMSGIYWGLTVMDLMGQLHRMNREEILAFIKSCQHECGGIS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5876
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.