Learn More
RABGGTB Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304323AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
RABGGTB catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.Specifications
RABGGTB | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P53611, P53612 | |
RABGGTB | |
Between 281 and 331 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RABGGTB | |
GGTB, GGTase-II-beta, geranylgeranyl transferase type II subunit beta, geranylgeranyl transferase type-2 subunit beta, rab GG transferase beta, rab GGTase beta, rab geranyl-geranyltransferase subunit beta, rab geranylgeranyltransferase subunit beta, type II protein ge | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19352, 5876 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.