Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabies Virus Antibody (Rab-50) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10063045
Description
Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Rabies Virus | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Clone Rab 50 shows conformation dependent specificity for the viral envelope glycoprotein. Not known to react with any other organism. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Rab-50 | |
| Unconjugated | |
| Glycoprotein | |
| Purified virions of SAD-Vnukova strain followed by Pitman-Moore strain. | |
| 0.5 mg | |
| Primary | |
| Virus | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction