Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabies Virus Antibody (Rab-50), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063045C
Description
Rabies Virus Monoclonal antibody specifically detects Rabies Virus in Virus samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Rabies Virus | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Rab-50 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Glycoprotein | |
Purified virions of SAD-Vnukova strain followed by Pitman-Moore strain. | |
0.5 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction