Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabies Virus Monoclonal Antibody (RAB-50)
GREENER_CHOICE

Catalog No. 01672082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-082 Supplier Invitrogen™ Supplier No. MA135778
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAb-50 reacts with SAD-Vnukovo and Pitman-Moore strains of rabies virus. It shows conformation dependent specificity for the viral envelope glycoprotein. In immunofluorescence test, SAD-Vnukovo strain is recognized by the antibody, while in Western blot, both strains are recognized by the antibody. In ELISA only SAD-Vnukovo strain is recognized. RAb-50 can neutralize the CVS strain of rabies virus.

Rabies virus (Neurotropic lyssavirus) is a member of the Rhabdoviridae family. Rabies is a single stranded, neurotropic, negative sense RNA virus which encodes 5 proteins: a glycoprotein, a nucleoprotein, and three others. The mature virus has a bullet shape, a protein coat, and a lipid envelope. The outer surface of the virus is covered with thumblike glycoprotein projections 5-10 nm long and 3 nm in diameter. The virus averages approximately 780 nm in length. Virus infectivity is destroyed by lipid solvents. Rabies virus is a very successful virus, with a very wide range of hosts. It causes an acute, central nervous system infection, characterized by CNS irritation, followed by paralysis and death. Approximately 50,000 human deaths each year are caused by rabies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus
Applications ELISA, Immunohistochemistry (Frozen), Neutralization, Western Blot
Classification Monoclonal
Clone RAB-50
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Neurotropic lyssavirus
Host Species Mouse
Immunogen Purified virions of Rabies virus, first of SAD-Vnukova strain, followed by Pitman-Moore strain.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.