Learn More
Invitrogen™ Rabies Virus Monoclonal Antibody (RV1C5)

Description
MA1-90618 detects Rabies virus from virus samples.
Specifications
Specifications
| Antigen | Rabies Virus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | RV1C5 |
| Concentration | 4.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Neurotropic lyssavirus |
| Host Species | Mouse |
| Immunogen | Purified rabies virus. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.