Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabies Virus Mouse anti-A Broad Range Of Species, Clone: 1112-1, MilliporeSigma™

Catalog No. MABF949MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF949MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF949MI Supplier MilliporeSigma Supplier No. MABF949
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rabies Virus, clone 1112-1 is a mouse monoclonal antibody that detects glycoprotein of strain ERA rabbies virus. It has been tested for use in Flow Cytometry and for Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone 1112-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Glycoprotein of ERA (Evelyn Rokitniki Abelseth) strain Rabies virus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 1112-1 detects glycoprotein of ERA strain of rabies virus in L929 cells that express rabies protein.
Target Species All species
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.