Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabies Virus Antibody (1C5cc), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211630R0.2ML
Description
Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Rabies Virus | |
Monoclonal | |
DyLight 550 | |
50 mM Sodium Borate | |
It has a virus-neutralizing activity in respect to the majority of standard RV and rabies-relative viruses: CVS, Lagos bat, Mokola, Flury lep, Duvenhage and the field isolates | |
Virus | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1C5 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Mouse | |
0.2 mL | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction