Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabies Virus Mouse anti-Virus, Unconjugated, Clone: , MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF94925UG
Description
Anti-Rabies Virus, clone 1112-1, Cat. No. MABF949, is a mouse monoclonal antibody that detects glycoprotein of strain ERA rabbies virus. It has been tested for use in Flow Cytometry and for Neutralizing.
Specifications
Rabies Virus | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Virus | |
Antibodies | |
IgG1 |
Flow Cytometry, Neutralization | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
Glycoprotein of ERA (Evelyn Rokitniki Abelseth) strain Rabies virus. | |
25 μg | |
Inflammation & Immunology | |
Clone 1112-1 detects glycoprotein of ERA strain of rabies virus in L929 cells that express rabies protein. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction