Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabies virus Mouse anti-Virus, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF203825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2038-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2038-25 Supplier MilliporeSigma Supplier No. MABF203825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Rabies virus, clone 802, Cat. No. MABF2038, is a mouse monoclonal antibody that detects glycoprotein of Rabies virus and has been tested for use in Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies virus
Applications Immunohistochemistry
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS without preservatives.
Gene Alias Glycoprotein G Rabies Virus
Host Species Mouse
Immunogen Beta-propiolactone-inactivated Rabies Virus (ERA strain).
Molecular Weight of Antigen 58.6 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 802 detects Glycoprotein of Rabies virus (strain ERA). This clone does not display neutralizing activity in vitro.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.