Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabies Virus, Serum Free Mouse anti-Virus, Unconjugated, Clone: 1112-1, MilliporeSigma™

Catalog No. MABF949SF10 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF949-SF-10 100 μg
MABF949-SF-25 25 μg
MABF949-SF-50 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MABF949-SF-10 Supplier MilliporeSigma Supplier No. MABF949SF100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rabies Virus, Serum Free, clone 1112-1, Cat. No. MABF949-SF, is a mouse monoclonal antibody that detects Glycoprotein of rabies virus., strain ERA and has been tested for use in ELISA, Flow Cytometry, and Neutralizing applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabies Virus, Serum Free
Applications ELISA, Flow Cytometry, Neutralization
Classification Monoclonal
Clone 1112-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Glycoprotein;Glycoprotein Rabies virus
Host Species Mouse
Immunogen Protein G of Evelyn Rokitniki Abelseth(ERA) SAD derived RABV strain).
Molecular Weight of Antigen 58.66 kDa calculated.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NA
Test Specificity Clone 1112-1 is a mouse monoclonal antibody that detects detects glycoprotein of strain ERA rabbies virus.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.