Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABIF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584226 Supplier Invitrogen™ Supplier No. PA584226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDLLQEHWLV EDMFIFENVG FTKDVGNIKF LVCADCEIGP IGWHCLDDKN SFYVALERVS HE Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 97%.

The Sec4/Rab-related small GTP-binding proteins are involved in the regulation of intracellular vesicular transport. Mss4 stimulates GTP-GDP exchange in Sec4 and Rab and binds to a subset of genetically related Rab proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABIF
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RABIF
Gene Accession No. P47224
Gene Alias AI842864; AW549708; E130318E12Rik; Guanine nucleotide exchange factor MSS4; guanine-nucleotide-releasing protein mss4; mammalian suppressor of SEC4; Mss4; RAB interacting factor; RABIF; RAB-interacting factor; RASGFR3; RASGRF3; Ras-specific guanine-releasing factor 3
Gene Symbols RABIF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RABIF. Recombinant protein control fragment (Product #RP-108406).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5877
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.