Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABIF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120327 Supplier Invitrogen™ Supplier No. PA5120327
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: DU145, mouse testis, rat brain. Immunogen sequence: MEPAEQPSEL VSAEGRNRKA VLCQRCGSRV LQPGTALFSR RQLFLPSMRK KPALSDGSNP DGDLLQEHWL VEDMFIFENV GFTKDVGNIK FLVCADCEIG PIGWHCLDDK NSFYVALERV SHE.

The Sec4/Rab-related small GTP-binding proteins are involved in the regulation of intracellular vesicular transport. Mss4 stimulates GTP-GDP exchange in Sec4 and Rab and binds to a subset of genetically related Rab proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABIF
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RABIF
Gene Accession No. P47224, Q08326, Q91X96
Gene Alias AI842864; AW549708; E130318E12Rik; Guanine nucleotide exchange factor MSS4; guanine-nucleotide-releasing protein mss4; mammalian suppressor of SEC4; Mss4; RAB interacting factor; RABIF; RAB-interacting factor; RASGFR3; RASGRF3; Ras-specific guanine-releasing factor 3
Gene Symbols RABIF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human RABIF (NP_002862.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304807, 5877, 98710
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.