Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABL2A Antibody (OTI4A8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273776PE
Description
RABL2A Monoclonal specifically detects RABL2A in Human, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
RABL2A | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
RABL2A | |
Full length recombinant protein of human RABL2A (NP_009013) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Monkey, Primate | |
IgG1 |
Flow Cytometry | |
OTI4A8 | |
Flow Cytometry | |
FLJ78724, MGC117180, RAB, member of RAS oncogene family-like 2A, rab-like protein 2A | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
11159.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction