Learn More
Invitrogen™ RABL2A Monoclonal Antibody (OTI4A8)

Description
RABL2A Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | RABL2A |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4A8 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RABL2A |
| Gene Accession No. | Q9UBK7 |
| Gene Alias | 1110031N17Rik; AW549303; RAB, member of RAS oncogene family-like 2; RAB, member of RAS oncogene family-like 2A; RAB, member of RAS oncogene family-like 2B; RAB, member RAS oncogene family-like 2; Rabl2; RABL2A; Rabl2b; rab-like protein 2A; rab-like protein 2B; RP11-395L14.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.