Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABL2A Monoclonal Antibody (OTI5A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502023S


Description
RABL2A Monoclonal antibody specifically detects RABL2A in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
RABL2A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UBK7 | |
Mouse | |
Affinity chromatography | |
RUO | |
11159, 362987 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5A10 | |
Unconjugated | |
RABL2A | |
1110031N17Rik; AW549303; RAB, member of RAS oncogene family-like 2; RAB, member of RAS oncogene family-like 2A; RAB, member of RAS oncogene family-like 2B; RAB, member RAS oncogene family-like 2; Rabl2; RABL2A; Rabl2b; rab-like protein 2A; rab-like protein 2B; RP11-395L14.2 | |
Full length human recombinant protein of human RABL2A produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction