Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABL2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560584
Description
Immunogen sequence: DKTKPSELDQ GKYDADDNVK IICLGDSAVG KSKLMERFLM DGFQPQQLST YALTLYKHTA TV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 89%.
This gene is a member of the RAB gene family which belongs to the RAS GTPase superfamily. The proteins in the family of RAS-related signaling molecules are small GTP-binding proteins that play important roles in the regulation of exocytotic and endocytotic pathways. This gene maps to the site of an ancestral telomere fusion event and may be a subtelomeric gene. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
RABL2A | |
Polyclonal | |
Unconjugated | |
RABL2A | |
1110031N17Rik; AW549303; RAB, member of RAS oncogene family-like 2; RAB, member of RAS oncogene family-like 2A; RAB, member of RAS oncogene family-like 2B; RAB, member RAS oncogene family-like 2; Rabl2; RABL2A; Rabl2b; rab-like protein 2A; rab-like protein 2B; RP11-395L14.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBK7 | |
RABL2A | |
Recombinant protein corresponding to Human RABL2A. Recombinant protein control fragment (Product #RP-104364). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction