Learn More
RABL2B Monoclonal Antibody (1B10)

Description
RABL2B Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | RABL2B |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1B10 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RABL2B |
| Gene Accession No. | Q9UNT1 |
| Gene Alias | HOX1B; RAB, member of RAS oncogene family-like 2B; RABL2B; rab-like protein 2B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.