Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557178
Description
Immunogen sequence: LAEDFNPEEI NLDCTNPRYL AAGSSNAVKL SRFFDKVIEK RYFLREGNQI PGFPDRKRFG AGTLKSLHYD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Predicted to enable GTP binding activity; GTPase activity; and protein homodimerization activity. Involved in regulation of Ras protein signal transduction and regulation of protein lipidation. Predicted to be active in endomembrane system. Implicated in pancreatic cancer.
Specifications
RABL3 | |
Polyclonal | |
Unconjugated | |
RABL3 | |
4930553C05Rik; AA958834; RAB, member of RAS oncogene family like 3; RAB, member of RAS oncogene family-like 3; RAB, member RAS oncogene family-like 3; Rabl3; rab-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285282 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5HYI8 | |
RABL3 | |
Recombinant protein corresponding to Human RABL3. Recombinant protein control fragment (Product #RP-96363). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction