Learn More
Invitrogen™ RABL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589757
Description
Immunogen sequence: MASLDRVKVL VLGDSGVGKS SLVHLLCQNQ VLGNPSWTVG CSVDVRVHDY KEGTPEEKTY YIELWDVGGS VGSASSVKST RAVFYNSVNG IIFVHDLTNK KSSQNLRRWS LEALNRDLVP TGVLVTNGDY DQEQFADNQI PLLVIGTKLD QIHETKRHEV LTRTAFLAED FNPEEINLDC TNPRYLAAGS SNAVKLSRFF DKVIEKRYFL REGNQIPGFP DRKRFGAGTL KSLHYD; Positive Samples: MCF7, Mouse brain, Mouse lung, Mouse teatis.
Predicted to enable GTP binding activity; GTPase activity; and protein homodimerization activity. Involved in regulation of Ras protein signal transduction and regulation of protein lipidation. Predicted to be active in endomembrane system. Implicated in pancreatic cancer.
Specifications
RABL3 | |
Polyclonal | |
Unconjugated | |
RABL3 | |
4930553C05Rik; AA958834; RAB, member of RAS oncogene family like 3; RAB, member of RAS oncogene family-like 3; RAB, member RAS oncogene family-like 3; Rabl3; rab-like protein 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
285282, 67657 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.84 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q5HYI8, Q9D4V7 | |
RABL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-236 of human RABL3 (NP_7761862). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.