Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553795 Supplier Invitrogen™ Supplier No. PA553795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNEESFNNCS KWLEKARSQA PGISLPGVLV GNKTDLAGRR AVDSAEARAW ALGQGLECFE TSVKEMENFE APFHCLAKQF HQLYREKVEV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

RABL4 belongs to the small GTPase superfamily, Ras family. RABL4 possesses GTPase activity. This gene encodes a putative GTP-binding protein similar to RAY/RAB1C. The protein is ras-related, but the function is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABL4
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT27
Gene Accession No. Q9BW83
Gene Alias 2600013G09Rik; BBS19; IFT27; intraflagellar transport 27; intraflagellar transport 27 homolog; intraflagellar transport protein 27 homolog; Putative GTP-binding protein RAY-like; RAB, member of RAS oncogene family-like 4; RABL4; rab-like protein 4; RAS oncogene family-like 4; RAYL
Gene Symbols IFT27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RABL4. Recombinant protein control fragment (Product #RP-92354).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.