Learn More
Invitrogen™ RABL4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total IFT27.
Specifications
Specifications
| Antigen | RABL4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | IFT27 |
| Gene Accession No. | Q9BW83, Q9D0P8 |
| Gene Alias | 2600013G09Rik; BBS19; IFT27; intraflagellar transport 27; intraflagellar transport 27 homolog; intraflagellar transport protein 27 homolog; Putative GTP-binding protein RAY-like; RAB, member of RAS oncogene family-like 4; RABL4; rab-like protein 4; RAS oncogene family-like 4; RAYL |
| Gene Symbols | IFT27 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.