Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560163
Description
Immunogen sequence: MLKAKILFVG PCESGKTVLA NFLTESSDIT EYSPTQGVRI LEFENPHVTS NNKGTGCEFE LWDCGGDAKF ESCWPALMKD AHGVVIV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Small GTPase-like component of the intraflagellar transport (IFT) complex B.
Specifications
RABL5 | |
Polyclonal | |
Unconjugated | |
Ift22 | |
3110017O03Rik; AI835745; AI847427; IFT22; intraflagellar transport 22; intraflagellar transport 22 homolog; intraflagellar transport protein 22 homolog; RAB, member of RAS oncogene family-like 5; RAB, member RAS oncogene family-like 5; Rabl5; rab-like protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H7X7 | |
Ift22 | |
Recombinant protein corresponding to Human RABL5. Recombinant protein control fragment (Product #RP-92713). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction