Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104052 Supplier Invitrogen™ Supplier No. PA5104052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RABL5.

Small GTPase-like component of the intraflagellar transport (IFT) complex B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABL5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ift22
Gene Accession No. Q5FVJ7, Q9H7X7
Gene Alias 3110017O03Rik; AI835745; AI847427; IFT22; intraflagellar transport 22; intraflagellar transport 22 homolog; intraflagellar transport protein 22 homolog; RAB, member of RAS oncogene family-like 5; RAB, member RAS oncogene family-like 5; Rabl5; rab-like protein 5
Gene Symbols Ift22
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFT22(Accession Q9H7X7), corresponding to amino acid residues E31-A81.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288585, 64792
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.