Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546208
Description
Peptide sequence: PFVEEYIPTQ EIQVTSIHWS YKTTDDIVKV EVWDVVDKGK CKKRGDGLKM Sequence homology: Cow: 100%; Dog: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 92%.
Maternal embryonic leucine-zipper kinase (MELK) is a key regulator of survival of stemlike GBM cells in vitro. MELK expression is increased in breast cancer tissue and this increase is also associated with poor patient survival, as predicted for a candidate oncogene.
Specifications
| RABL6 | |
| Polyclonal | |
| Unconjugated | |
| RABL6 | |
| B230208H17Rik; C11H9orf86; C9H9orf86; C9orf86; GTP-binding protein Parf; PARF; Partner of ARF; pp8875; putative GTP-binding protein Parf; RAB, member of RAS oncogene family-like 6; RAB, member RAS oncogene family like 6; RAB, member RAS oncogene family-like 6; RABL6; Rab-like GTP-binding protein 1; Rab-like GTP-binding protein 1A; Rab-like GTP-binding protein 1C; rab-like protein 1; Rab-like protein 6; RBEL1; Rbel1a; Rbel1b; RGD1307615 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55684 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3YEC7 | |
| RABL6 | |
| Synthetic peptide directed towards the N-terminal of human RABL6 (aa 67-116). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction