Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABV Glycoprotein Mouse anti-RabiesVirus, Unconjugated, Clone: MAb-D1, MilliporeSigma™

Catalog No. MABF19625MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF19625MI 25 μg
MABF1967BMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF19625MI Supplier MilliporeSigma Supplier No. MABF1967B25UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RABV Glycoprotein Monoclonal antibody specifically detects RABV Glycoprotein in RabiesVirus samples. It is validated for ELISA, Immunoprecipitation, Neutralization Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABV Glycoprotein
Applications ELISA, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone MAb-D1
Conjugate Unconjugated
Gene Accession No. NP_056796
Gene Alias Glycoprotein
Gene Symbols G
Host Species Mouse
Immunogen Inactivated and purified Rabies Virus (Pasteur virus strain).
Molecular Weight of Antigen 58.56 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone MAb-D1 is a mouse monoclonal antibody that detects Rabies virus glycoprotein. It targets an epitope within nine amino acids in the antigenic site III.
Target Species Rabies virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.