Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABV Glycoprotein, serum free Mouse anti-Virus, Clone: D1-25, MilliporeSigma™

Catalog No. MABF1967100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1967100 100 μg
MABF196725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1967100 Supplier MilliporeSigma Supplier No. MABF1967100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RABV Glycoprotein, serum free Monoclonal antibody specifically detects RABV Glycoprotein, serum free in Virus samples. It is validated for ELISA, Immunoprecipitation, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABV Glycoprotein, serum free
Applications ELISA, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone D1-25
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Host Species Mouse
Immunogen Inactivated and purified Rabies Virus (Pasteur virus strain).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.