Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ rac BHFF
GREENER_CHOICESDP

Catalog No. 331350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-135-0 50 mg
33-131-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-135-0 Supplier Tocris Bioscience™ Supplier No. 3313/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective postitive allosteric modulator of GABAB receptors

Potent and selective GABAB receptor positive allosteric modulator that increases the potency and efficacy of GABA ( > 15-fold and > 149% respectively). Exhibits anxiolytic activity in vivo and is orally active.

Chemical Identifiers

CAS 123557-91-5
Molecular Formula C17H21F3O3

Specifications

Quantity 50 mg
Formula Weight 330.34
Purity >98%
Molecular Weight (g/mol) 330.347
Chemical Name or Material rac BHFF
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.