Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAC1 Monoclonal Antibody (1301CT276.121.104)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537658
Description
RAC1 Monoclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
RAC1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P63000, P63001, Q6RUV5 | |
Rac1 | |
KLH conjugated synthetic peptide. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1301CT276.121.104 | |
Unconjugated | |
Rac1 | |
AL023026; cell migration-inducing gene 5 protein; D5Ertd559e; GTPase; MGC111543; MIG5; p21-Rac1; Rac 1b; Rac family small GTPase 1; RAC1; Rac-1; Rac2; rac2 GTP-binding protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; TC25; TC-25 | |
Mouse | |
Protein G | |
RUO | |
19353, 363875, 5879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction