Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rac1 Antibody (CPTC-RAC1-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308939
Description
Rac1 Monoclonal specifically detects Rac1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
Rac1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Immunohistochemistry-Paraffin, Protein Array | |
Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
Recombinant human full-length Rac1protein | |
100 μg | |
Breast Cancer, Cancer, mTOR Pathway, Phospho Specific | |
5879 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
CPTC-RAC1-1 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction