Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAC1/RAC2/RAC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561600 Supplier Invitrogen™ Supplier No. PA561600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKLDLRDDKD TIERLRDKKL APITYPQGLA MAREI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAC1/RAC2/RAC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAC2
Gene Accession No. P15153, P60763, P63000
Gene Alias AI323801; AI452260; AL023026; Cell migration-inducing gene 5 protein; D5Ertd559e; EN-7; EN-7 protein; GTPase; Gx; HSPC022; MIG5; p21-Rac1; p21-Rac2; p21-Rac3; protein EN-7; rac; Rac family small GTPase 1; Rac family small GTPase 1 L homeolog; Rac family small GTPase 2; Rac family small GTPase 3; rac1; Rac-1; rac1.L; Rac1B; RAC2; Rac3; Ras-like protein TC25; RAS-related C3 botulinum substrate 1; RAS-related C3 botulinum substrate 2; RAS-related C3 botulinum substrate 3; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) L homeolog; ras-related C3 botulinum toxin substrate 2; ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2); ras-related C3 botulinum toxin substrate 3; ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3); Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); rho family, small GTP binding protein Rac1; rho family, small GTP binding protein Rac3; small G protein; TC25; TC-25; XELAEV_18045321mg; Xrac
Gene Symbols Rac1, RAC2, Rac3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAC1/RAC2/RAC3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5879, 5880, 5881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.