Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rac2, Rabbit, Unlabeled, Polyclonal,

Catalog No. 07604I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07604I 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07604I Supplier MilliporeSigma™ Supplier No. 07604I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ras-related C3 botulinum toxin substrate 2 (UniProt P15153; also known as GX, p21-Rac2, Small G protein) is encoded by the RAC2 gene (Gene ID 5880) in human. Rho family of small GTPases are involved in a wide variety of cellular functions such as cell polarity, vesicular trafficking, and cell cycle by regulating intracellular actin dynamics. In mammals, the Rho family contains 22 members which can be further divided into the Rac subfamily (Rac1, Rac2, Rac3, and RhoG), Cdc42 subfamily (Cdc42, TC10, TCL, Chip, and Wrch-1), RhoA subfamlly (RhoA, RhoB, and RhoC) and other Rho-GTPases (RhoE/Rnd3, RhoH/TTF, Rif, RhoBTB1, RhoBTB2, Miro-1, Miro-2, RhoD, Rnd1, and Rnd2). Rac1 and Rac2 share high sequence homology (90%) and are both expressed in the hematopoietic system, where they play crucial roles in mediating the interactions between hematopoietic stem cells (HSCs) and their microenvironment. Rac proteins transduce cell surface signaling events initiated by tyrosine kinase receptors, G protein-coupled receptors, and cell-to-cell contacts by acting as molecular switches, cycling between an inactive GDP-bound state and a GTP-bound active state. In addition to overlapping functions, Rac1 and Rac2 also mediate unique functions via interactions with specific partners. SAM50 and Metaxin1, for example, are among mitochondrial proteins identified to be specifically associated with Rac2. Rac activity inhibition is reported to effectively target primary human AML and CML cells and prevent disease initiation in various murine leukemia models.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RAC2
Gene Accession No. P15153
Host Species Rabbit
Immunogen GST-tagged recombinant full-length human Rac2 pro-form.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002863
Test Specificity This rabbit polyclonal antibody was raised against recombinant full-length human Rac2 pro-form (a.a. 1-192) and therefore will recognize both the pro- and mature (a.a. 1-189) Rac2.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.