Learn More
Fisherbrand™ Racer Series Qualifier Eyewear
SCI_ED
Wraparound design offers superior side coverage.
Specifications
Lens Coating Scratch-resistant
Lens Material Polycarbonate
Frame Material Nylon
View More Specs
Products 3
Catalog Number Frame Color Lens Tint Price Quantity & Availability  
Catalog Number Frame Color Lens Tint Price Quantity & Availability  
S98423
SCI_ED
View Documents
Red Clear N/A N/A N/A
S98424
SCI_ED
View Documents
Red Dark Gray N/A N/A N/A
S98420
SCI_ED
View Documents
Black Clear N/A N/A N/A
Description
Description
  • Scratch-resistant, clear polycarbonate lenses block >99.9% of harmful UV radiation
  • Wraparound design offers superior side coverage
  • Lightweight frames are durable and comfortable
  • Flared single-lens design provides sleek protection
  • A soft nose bridge and soft, bayonet-style side arms grip the face and temples lightly but firmly for a comfortable fit
  • Choose from black or red nylon frames
Specifications
Specifications
Scratch-resistant
Polycarbonate
Nylon
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.