Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RACGAP1 Monoclonal Antibody (23A8)
GREENER_CHOICE

Catalog No. MA120580
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA120580 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA120580 Supplier Invitrogen™ Supplier No. MA120580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: rat brain microsomal protein. MA1-20580 detects a band of approximately 21 kDa.

RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RACGAP1
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 30% glycerol and 0.05% sodium azide
Gene RACGAP1
Gene Accession No. Q9H0H5, Q9WVM1
Gene Alias AI227039; AI327394; AKT; Band25; CYK4; GAP; gtl11; GTPase; HsCYK-4; ID; ID-GAP; KIAA1478; Male germ cell RacGap; MGC99656; MgcRacGAP; mKIAA1478; PKB; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase activating protein 1; Rac GTPase-activating protein 1; RAC-ALPHA; Racgap1
Gene Symbols RACGAP1
Host Species Mouse
Immunogen Recombinant protein containing the full length human Rac1.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26934, 29127, 315298
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.