Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RACGAP1 Mouse anti-Human, Mouse, Rat, Clone: 6E10A2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | RACGAP1 |
|---|---|
| Clone | 6E10A2 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6495
![]()
|
Proteintech Group Inc
660561IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Rho GTPases control a variety of cellular processes. There are 3 subtypes of Rho GTPases in the Ras superfamily of small G proteins: RHO (see MIM 165370), RAC (see RAC1; MIM 602048), and CDC42 (MIM 116952). GTPase-activating proteins (GAPs) bind activated forms of Rho GTPases and stimulate GTP hydrolysis. Through this catalytic function, Rho GAPs negatively regulate Rho-mediated signals. GAPs may also serve as effector molecules and play a role in signaling downstream of Rho and other Ras-like GTPases.Specifications
| RACGAP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HsCYK 4, ID GAP, KIAA1478, Male germ cell RacGap, MgcRacGAP, RACGAP1 | |
| Racgap1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 6E10A2 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9H0H5, Q9WVM1 | |
| 26934, 29127, 315298 | |
| RACGAP1 Fusion Protein Ag6944 | |
| Primary | |
| -20°C | |
| RACGAP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title