Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RACK1/GNB2L1 Antibody (OTI2D8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273781H
Description
RACK1/GNB2L1 Monoclonal specifically detects RACK1/GNB2L1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RACK1/GNB2L1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GNB2L1 | |
Full length recombinant protein of human RACK1/GNB2L1 (NP_006089) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D8 | |
Western Blot, Immunohistochemistry | |
guanine nucleotide binding, guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1, Receptor for activated C kinase, Receptor for Activated C Kinase 1 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
10399.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction