Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RACK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142671
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142671 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142671 Supplier Invitrogen™ Supplier No. PA5142671
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RACK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RACK1
Gene Accession No. P63244, P63245, P68040
Gene Alias 12-3; activated protein kinase C receptor; AL033335; Cell proliferation-inducing gene 21 protein; CG7111; CG7111-PA; CG7111-PB; CG7111-PC; CG7111-PD; Dmel\CG7111; Dmel_CG7111; dRack1; GB-like; gnb2l1; Gnb2-rs1; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; guanine nucleotide-binding protein beta-subunit like protein; guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein; Guanine nucleotide-binding protein subunit beta-like protein 12.3; H12.3; HLC7; HLC-7; Human lung cancer oncogene 7 protein; i173; lung cancer oncogene 7; OTTHUMP00000223870; OTTHUMP00000223891; OTTHUMP00000223893; OTTHUMP00000223900; OTTHUMP00000223901; OTTHUMP00000223902; OTTHUMP00000223930; OTTHUMP00000223931; p205; PIG21; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; RACK; Rack1; rack-1; Rack1-PA; Rack1-PB; Rack1-PC; Rack1-PD; RE74715p; receptor for activated C kinase; receptor for activated C kinase 1; receptor for activated protein kinase C; receptor for activated protein kinase C1; receptor of activated PKC; receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; Receptor of activated protein kinase C; receptor of activated protein kinase C 1; Receptor of activated protein kinase C homolog; similar to mammalian RACK1; Small ribosomal subunit protein RACK1; wu:fb80d08; wu:fk65d12
Gene Symbols RACK1
Host Species Goat
Immunogen Peptide with sequence NTLGVCKYTVQDESH.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10399, 14694, 83427
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.