Learn More
RACK1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302545AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Bovine and Pig
RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.Specifications
RACK1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P63244, P68040 | |
Rack1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RACK1 | |
Guanine nucleotide-binding protein subunit beta-like protein 12.3, Receptor of activated protein kinase C 1, RACK1, Receptor for activated C kinase, Guanine nucleotide-binding protein subunit beta-2-like 1, GNB2L1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10399, 14694 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.